missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 2.00 Normal, Aqueous, Reagents
Perchloric Acid, 2.00 Normal, Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CP121300500C
Specifications
| Perchloric Acid, 2.00 Normal, Aqueous | |
| 7732-18-5,7601-90-3 | |
| Glass Bottle | |
| 500 mL | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
| Colorless | |
| 101°C | |
| 1.1145 | |
| H2O | |
| Chlorine Tetraoxide | |
| O | |
| 18.02 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Chlorine Tetraoxide | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |