missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 2.00 N Aqueous, Ricca Chemical

Catalog No. R55167001C
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R55167001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R55167001C Supplier Ricca Chemical Company Supplier No. R55167001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Color Colorless
Concentration 2.00 N
CAS Min % 76.0, 21.71
CAS Max % 79.0, 22.59
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Perchloric Acid