missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.30 Normal, in P-Dioxane, Reagents

Catalog No. 01334298
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334298 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334298 Supplier Labchem Inc Supplier No. CP1207004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perchloric Acid, 0.30 Normal, in P-Dioxane, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 123-91-1 , 7601-90-3 , 7732-18-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide in 1,4-Dioxane
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Color Colorless
Boiling Point 101°C
Packaging Glass Bottle
Specific Gravity 1.0503
Quantity 4 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.30 Normal, in P-Dioxane