missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.2% v/v, Reagents

Perchloric Acid, 0.2% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$270.57 - $270.57

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide
IUPAC Name water
SMILES O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-334-293
View Documents
Labchem Inc
CP1211004A
4 L N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Chlorine Tetraoxide
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

<2
7732-18-5,7601-90-3
Poly Bottle
4 L
Liquid
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
0.2%
Perchloric Acid, 0.2% v/v
Colorless
100°C
1.0013
H2O
Chlorine Tetraoxide
O
18.02
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'