missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.2% v/v, Reagents

Catalog No. 01334293
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334293 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334293 Supplier Labchem Inc Supplier No. CP1211004A

Perchloric Acid, 0.2% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7601-90-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide
IUPAC Name water
SMILES O

Specifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0013
Quantity 4 L
Physical Form Liquid
Percent Purity 0.2%
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.2% v/v