missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane, Reagents

Catalog No. p-200021849
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334310 1 L
01334309 4 L
01334308 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334310 Supplier Labchem Inc Supplier No. CP1202001C
Only null left

Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 123-91-1 , 7601-90-3 , 7732-18-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide in 1,4-Dioxane
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Color Colorless
Boiling Point 103°C
Packaging Glass Bottle
Specific Gravity 1.0394
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane