missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.1% Made with HPLC Water, Reagents

Catalog No. 01334288
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334288 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334288 Supplier Labchem Inc Supplier No. CP1214001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perchloric Acid, 0.1% Made with HPLC Water, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7601-90-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide
IUPAC Name water
SMILES O

Specifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.0007
Quantity 1 L
Physical Form Liquid
Percent Purity 0.1%
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.1% Made with HPLC Water