missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.055 N in n-Propanol, Reagents

Catalog No. 01334312
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334312 1 L
01334311 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334312 Supplier Labchem Inc Supplier No. CP1201001C
Only null left

Perchloric Acid, 0.055 N in n-Propanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-23-8 , 7601-90-3 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide in 1-Propanol
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Color Colorless
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.05
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.055 N in n-Propanol