missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 0.020 Normal (N/50) in Acetic Acid, Reagents
Perchloric Acid, 0.020 Normal (N/50) in Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CP1212004A
Specifications
| <2 | |
| Colorless to Light Yellow | |
| 118°C | |
| 1.0507 | |
| C2H4O2 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| Reagent |
| Perchloric Acid, 0.020 Normal (N/50) in Acetic Acid | |
| 64-19-7,7601-90-3,7732-18-5 | |
| Poly Bottle | |
| 4 L | |
| Liquid | |
| Chlorine Tetraoxide in Glacial Acetic Acid | |
| CC(O)=O | |
| 60.05 |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| Chlorine Tetraoxide in Glacial Acetic Acid | |
| CC(O)=O |