missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepstatin A, Fisher BioReagents
Supplier: Fisher BioReagents BP267110
Description
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.Specifications
| Pepstatin A | |
| 235°C | |
| 230° to 240°C | |
| Amber Glass | |
| 10 mg | |
| pepstatin a | |
| CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O | |
| 685.904 | |
| 685.9 |
| 26305-03-3 | |
| White | |
| Conforms to reference | |
| C34H63N5O9 | |
| 15, 7260 | |
| FAXGPCHRFPCXOO-WQILDUGRSA-N | |
| (3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid | |
| 131801262 | |
| Powder or Solid |
Chemical Identifiers
| 26305-03-3 | |
| 685.904 | |
| pepstatin a | |
| (3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid |
| C34H63N5O9 | |
| FAXGPCHRFPCXOO-WQILDUGRSA-N | |
| 131801262 | |
| CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O |
Safety and Handling
Recommended Storage : RT