missing translation for 'onlineSavingsMsg'
Learn More

Pepstatin A, Fisher BioReagents

Catalog No. p-27522
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
250 mg
Packaging:
Amber Glass
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP26715 5 mg Amber Glass
BP267110 10 mg Amber Glass
BP267125 25 mg Amber Glass
BP2671250 250 mg Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP26715 Supplier Fisher BioReagents Supplier No. BP26715

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.

Chemical Identifiers

CAS 26305-03-3
Molecular Formula C34H63N5O9
Molecular Weight (g/mol) 685.904
InChI Key FAXGPCHRFPCXOO-WQILDUGRSA-N
Synonym pepstatin a
PubChem CID 131801262
IUPAC Name (3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid
SMILES CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O

Specifications

Melting Point 235°C
Color White
Melting Range 230° to 240°C
Infrared Spectrum Conforms to reference
Packaging Amber Glass
Quantity 5 mg
Merck Index 15, 7260
Formula Weight 685.9
Physical Form Powder or Solid
Chemical Name or Material Pepstatin A
Safety handling Check Alert storage code Red
Recommended Storage 0°C