Learn More
Gibco™ PeproGMP™ Human VEGF-165 Recombinant Protein, PeproTech®
Description
PeproTech Recombinant Human VEGF-165 is a 38.2 kDa, disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains. PeproGMP VEGF-165 can be used with PeproGMP BMP-4 and PeproGMP SCF in CTS StemScale medium for pluripotent stem cell differentiation, specifically for mesoderm and hematopoetic progenitor cell induction. Reconstitute using sterile water (i.e., WFI) to a concentration of 0.1-1.0 mg/ml. Additional product information, specifications, handling and reconstitution information are provided on the Certificate of Analysis. All PeproGMP™ products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter <1043> Ancillary Materials for Cell, Gene, and Tissue-Engineered Products.
Specifications
Specifications
| Accession Number | P15692 |
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 7422 |
| Molecular Weight (g/mol) | 38.2 kDa |
| Name | PeproGMP™ Human VEGF-165 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Storage Requirements | -20°C or -80°C if preferred |
| Endotoxin Concentration | ≤0.1 EU/μg |
| Show More |