Learn More
Gibco™ PeproGMP® Human IL-7 Recombinant Protein, PeproTech®
Recombinant Protein
$1868.00 - $1868.00
Specifications
| Accession Number | P13232, P16871 |
|---|---|
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Format | Lyophilized |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3574, 3575 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-672-622
|
Gibco™
GMP2000750UG |
50 μg |
Each for $1,868.00
|
|
|||||
Description
Recombinant Human IL-7 is a 17.4 kDa protein containing 152 amino acid residues. Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.Specifications
| P13232, P16871 | |
| Lyophilized | |
| 3574, 3575 | |
| PeproGMP™ Human IL-7 | |
| ≤0.1 EU/μg | |
| IL-7 | |
| Unconjugated | |
| DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |
| Control, Western Blot Control, Bioactivity | |
| Protein with no preservative | |
| 17.4 kDa | |
| GMP | |
| A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
| Determined by the proliferation of mouse 2E8 cells. PeproGMP Recombinant Human IL-7 ha been tested against the WHO standard NIBSC 90/530. | |
| Recombinant | |
| -20°Cor to +80°C if preferred | |
| Lyophilized |