missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human EGF Recombinant Protein, PeproTech®

Catalog No. p-200087557
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01672630 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01672630 Supplier Gibco™ Supplier No. GMP10015100UG
Only null left

Recombinant Protein

Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.

Specifications

Accession Number P01133
For Use With (Application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 1950
Molecular Weight (g/mol) 6.2 kDa
Name PeproGMP™ Human EGF
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status GMP
Endotoxin Concentration ≤0.1 EU/μg
Gene Alias AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone
Common Name EGF
Gene Symbol EGF
Biological Activity Determined by the proliferation of mouse Balb/c 3T3 cells. PeproGMP Recombinant Human EGF has been tested against the WHO standard NIBSC 91/530.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
Content And Storage -20°Cor to +80°C if preferred
Activity Determined by the proliferation of mouse Balb/c 3T3 cells. PeproGMP Recombinant Human EGF has been tested against the WHO standard NIBSC 91/530. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less