missing translation for 'onlineSavingsMsg'
Learn More

Peppermint Oil, NF, 5%, Spectrum™ Chemical

PE105, 8006-90-4

$426.03 - $1385.94

Chemical Identifiers

CAS 8006-90-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605010
View Documents
Spectrum Chemical Mfg Cor
PE105100ML
100 mL Amber Glass Bottle
Each for $426.03
Only null left
 
18605011
View Documents
Spectrum Chemical Mfg Cor
PE105500MLGL
500 mL Amber Glass Bottle
Each for $1,385.94
Only null left
 
Description

Description

Spectrum™ Chemical Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

8006-90-4

Specifications

8006-90-4
Amber Glass Bottle
100 mL
NF
1
1.459-1.465
5%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'