missing translation for 'onlineSavingsMsg'
Learn More

Peppermint Oil, NF, 5%, Spectrum™ Chemical

Catalog No. 18605011
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605011 500 mL Amber Glass Bottle
18605010 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605011 Supplier Spectrum Chemical Mfg Cor Supplier No. PE105500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PE105, 8006-90-4

Spectrum™ Chemical Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8006-90-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.459-1.465
Quantity 500 mL
Percent Purity 5%
Grade NF
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Peppermint Oil