missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
$1529.67 - $1529.67
Specifications
| For Use With | PepMap™ Neo Trap Cartridge (part number 174500) |
|---|---|
| Max. Pressure | 22,000 psi (1500 bar) |
| Temperature | 60°C |
| Product Line | PepMap Neo |
| For Use With (Application) | Trap and Elute, Peptide Mapping |
| Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
|
03-255-020
|
Thermo Scientific™
174502 |
Cartridge Holder Kit | Cartridge Holder, Tubing, Fittings |
Each for $1,529.67
|
|
||||
Description
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
See Viper and nanoViper resources
Specifications
| PepMap™ Neo Trap Cartridge (part number 174500) | |
| 60°C | |
| Trap and Elute, Peptide Mapping |
| 22,000 psi (1500 bar) | |
| PepMap Neo |