missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Description
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
Specifications
Specifications
| Diameter (Metric) | 30 μm (tubing) |
| For Use With | PepMap™ Neo Trap Cartridge (part number 174500) |
| Includes | Cartridge Holder, Tubing, Fittings |
| Length (Metric) | 5 mm |
| Max. Pressure | 22,000 psi (1500 bar) |
| Quantity | Each |
| Temperature | 60°C |
| Product Line | PepMap Neo |
| Description | Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings |
| For Use With (Application) | Trap and Elute, Peptide Mapping |
| Show More |