missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peonidin 3-O-glucoside chloride, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| CAS | 6906-39-4 |
| Physical Form | Neat |
| Linear Formula | C22H23 ClO11 |
| MDL Number | MFCD00189435 |
| Synonym | 3-(Glucosyloxy)-4 ′,5,7-trihydroxy-3 ′-methoxyflavylium chloride; Glucopeonidin chloride |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 498.86 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Analytical Standard |