missing translation for 'onlineSavingsMsg'
Learn More

Pentatriacontane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014973
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014973 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014973 Supplier Merck Emd Millipore Supplier No. 76968250MG
Petroleum

Specifications

CAS 630-07-9
Physical Form Neat
Linear Formula CH3(CH2)33 CH3
MDL Number MFCD00059397
Melting Point 73°C to 75°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 492.95
Percent Purity ≥98.0% (GC)
Grade Analytical Standard