missing translation for 'onlineSavingsMsg'
Learn More

Pentane, ≥99.5% (GC), MilliporeSigma™ Supelco™

$305.90 - $1004.09

Specifications

Ignition Point 500°F
CAS 109-66-0
Vapor Density 2.48 (vs. air)
Density 0.626 g/mL (at 25°C)
Flash Point -40°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019002
View Documents
Merck Emd Millipore
7687010ML
10 mL
Each for $305.90
Only null left
 
111018166
View Documents
Merck Emd Millipore
7687050ML
50 mL
Each for $1,004.09
Only null left
 
Description

Description

Petroleum
Specifications

Specifications

500°F
2.48 (vs. air)
-40°C
CH3(CH2)3 CH3
-130°C
Analytical Standard
≥99.5% (GC)
109-66-0
0.626 g/mL (at 25°C)
Neat
MFCD00009498
35°C to 36°C
72.15
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'