missing translation for 'onlineSavingsMsg'
Learn More

Pentamethyldisiloxane, 95%

CAS: 1438-82-0 | C5H15OSi2 | 147.344 g/mol

Supplier:  Thermo Scientific Chemicals L1657714

 View more versions of this product

Catalog No. AAL1657714


Only null left
Explore available promotions

Description

Description

Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents.
Specifications

Specifications

86°C to 87°C
1438-82-0
C5H15OSi2
UN1993
−19°C (−2°F)
Not miscible or difficult to mix in water.
C[Si](C)O[Si](C)(C)C
147.344
148.35
1.374
0.758
Pentamethyldisiloxane
25 g
MFCD00053658
1734568
pentamethyldisiloxane, disiloxane, pentamethyl, 1,1,1,3,3-pentamethyldisiloxane, dimethyl trimethylsilyloxy silicon, disiloxane, 1,1,1,3,3-pentamethyl, dimethyl trimethylsilyl oxy silyl, dimethylsilyl oxy trimethylsilane
XUKFPAQLGOOCNJ-UHFFFAOYSA-N
dimethyl(trimethylsilyloxy)silicon
6327260
95%
Moisture sensitive

Chemical Identifiers

1438-82-0
147.344
XUKFPAQLGOOCNJ-UHFFFAOYSA-N
6327260
C[Si](C)O[Si](C)(C)C
C5H15OSi2
MFCD00053658
pentamethyldisiloxane, disiloxane, pentamethyl, 1,1,1,3,3-pentamethyldisiloxane, dimethyl trimethylsilyloxy silicon, disiloxane, 1,1,1,3,3-pentamethyl, dimethyl trimethylsilyl oxy silyl, dimethylsilyl oxy trimethylsilane
dimethyl(trimethylsilyloxy)silicon
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 215-873-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only