missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentamethyldisiloxane, 95%
Description
Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.
Solubility
Not miscible or difficult to mix in water.
Notes
It is sensitive to moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents.
Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.
Solubility
Not miscible or difficult to mix in water.
Notes
It is sensitive to moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents.
Specifications
Specifications
| Boiling Point | 86°C to 87°C |
| Quantity | 5 g |
| UN Number | UN1993 |
| Beilstein | 1734568 |
| Flash Point | −19°C (−2°F) |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 148.35 |
| Percent Purity | 95% |
| Refractive Index | 1.374 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 215-873-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only