missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylbenzene 99.0+%, TCI America™

Catalog No. p-7134389
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00435G 5 g
P004325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00435G Supplier TCI America Supplier No. P00435G
Only null left

Chemical Identifiers

CAS 700-12-9
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008522
InChI Key BEZDDPMMPIDMGJ-UHFFFAOYSA-N
Synonym pentamethylbenzene, benzene, pentamethyl, pentamethylbenzol, benzene, 1,2,3,4,5-pentamethyl, pentamethyl phenyl, acmc-1bkt8, ksc492e3d, 1,3,4,5-pentamethylbenzene, pentamethylbenzene 5g
PubChem CID 12784
ChEBI CHEBI:38998
IUPAC Name 1,2,3,4,5-pentamethylbenzene
SMILES CC1=CC(=C(C(=C1C)C)C)C

Specifications

Melting Point 53°C
Color White
Boiling Point 231°C
Quantity 5 g
UN Number 1325
Formula Weight 148.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentamethylbenzene