missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylbenzene, 99%

CAS: 700-12-9 | C11H16 | 148.249 g/mol

Supplier:  Thermo Scientific Chemicals L0416322

 View more versions of this product

Catalog No. AAL0416322


Only null left
Explore available promotions

Description

Description

Pentamethylbenzene is used to prepare a mixture of nitropentamethylbenzene and 2,3,4,5-tetramethylbenzyl nitrate1. It was also used to prepare propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentamethylbenzene is used to prepare a mixture of nitropentamethylbenzene and 2,3,4,5-tetramethylbenzyl nitrate1. It was also used to prepare propene.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Pentamethylbenzene
50°C to 51°C
231°C to 232°C
C11H16
100 g
1905349
BEZDDPMMPIDMGJ-UHFFFAOYSA-N
1,2,3,4,5-pentamethylbenzene
12784
148.25
700-12-9
0.917
91°C (195°F)
MFCD00008522
UN1325
pentamethylbenzene, benzene, pentamethyl, pentamethylbenzol, benzene, 1,2,3,4,5-pentamethyl, pentamethyl phenyl, acmc-1bkt8, ksc492e3d, 1,3,4,5-pentamethylbenzene, pentamethylbenzene 5g
CC1=CC(=C(C(=C1C)C)C)C
148.249
CHEBI:38998
99%

Chemical Identifiers

700-12-9
148.249
BEZDDPMMPIDMGJ-UHFFFAOYSA-N
12784
1,2,3,4,5-pentamethylbenzene
C11H16
MFCD00008522
pentamethylbenzene, benzene, pentamethyl, pentamethylbenzol, benzene, 1,2,3,4,5-pentamethyl, pentamethyl phenyl, acmc-1bkt8, ksc492e3d, 1,3,4,5-pentamethylbenzene, pentamethylbenzene 5g
CHEBI:38998
CC1=CC(=C(C(=C1C)C)C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 211-837-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only