missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylbenzaldehyde 96.0+%, TCI America™

Catalog No. P13511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13511G 1 g
P13515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13511G Supplier TCI America Supplier No. P13511G
Only null left

Chemical Identifiers

CAS 17432-38-1
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00015560
InChI Key RWOZGGOKRKSHKN-UHFFFAOYSA-N
Synonym pentamethylbenzaldehyde, pubchem16352, acmc-1busy, 3,4-dibenzyloxyphenethylamine hydroxide
PubChem CID 263712
IUPAC Name 2,3,4,5,6-pentamethylbenzaldehyde
SMILES CC1=C(C(=C(C(=C1C)C)C=O)C)C

Specifications

Melting Point 150°C
Quantity 1 g
Formula Weight 176.26
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentamethylbenzaldehyde