missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic Anhydride 95.0+%, TCI America™

Supplier:  TCI America P056625G

 View more versions of this product

Catalog No. P056625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pentafluoropropionic Anhydride
Yellow
C6F10O3
25 g
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F
310.047
310.05
Liquid
356-42-3
80°C
MFCD00000429
1760
XETRHNFRKCNWAJ-UHFFFAOYSA-N
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
67742
≥95.0% (GC)

Chemical Identifiers

356-42-3
310.047
XETRHNFRKCNWAJ-UHFFFAOYSA-N
67742
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F
C6F10O3
MFCD00000429
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions