missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenylhydrazine 98.0+%, TCI America™

Catalog No. p-7134810
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08525G 5 g
P085225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08525G Supplier TCI America Supplier No. P08525G
Only null left

Chemical Identifiers

CAS 828-73-9
Molecular Formula C6H3F5N2
Molecular Weight (g/mol) 198.096
MDL Number MFCD00007574
InChI Key BYCUWCJUPSUFBX-UHFFFAOYSA-N
Synonym pentafluorophenylhydrazine, pentafluorophenyl hydrazine, perfluorophenyl hydrazine, hydrazine, pentafluorophenyl, 2,3,4,5,6-pentafluorophenyl hydrazine, 1-2,3,4,5,6-pentafluorophenyl hydrazine, pubchem3316, pentafluorophenyl-hydrazine
PubChem CID 13236
IUPAC Name (2,3,4,5,6-pentafluorophenyl)hydrazine
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)NN

Specifications

Melting Point 76°C
Quantity 5 g
Formula Weight 198.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorophenylhydrazine
RTECSNumber MV8325000
TSCA No
Recommended Storage Refrigerator