missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyldimethylchlorosilane 95.0+%, TCI America™
$210.63 - $822.05
Chemical Identifiers
| CAS | 20082-71-7 |
|---|---|
| Molecular Formula | C8H6ClF5Si |
| Molecular Weight (g/mol) | 260.66 |
| MDL Number | MFCD00000500 |
| InChI Key | PQRFRTCWNCVQHI-UHFFFAOYSA-N |
| Synonym | pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane |
| PubChem CID | 88361 |
| IUPAC Name | chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane |
| SMILES | C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F |
Chemical Identifiers
| 20082-71-7 | |
| 260.66 | |
| PQRFRTCWNCVQHI-UHFFFAOYSA-N | |
| 88361 | |
| C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F |
| C8H6ClF5Si | |
| MFCD00000500 | |
| pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane | |
| chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane |
Specifications
| 20082-71-7 | |
| 47°C | |
| MFCD00000500 | |
| 2985 | |
| PQRFRTCWNCVQHI-UHFFFAOYSA-N | |
| chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane | |
| 88361 | |
| ≥95.0% (GC) | |
| Pentafluorophenyldimethylchlorosilane [Pentafluorophenyldimethylsilylating Agent] |
| Yellow | |
| C8H6ClF5Si | |
| 1 mL | |
| pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane | |
| C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F | |
| 260.66 | |
| 260.66 | |
| Liquid |
Safety and Handling
TSCA : Yes