missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyldimethylchlorosilane 95.0+%, TCI America™

Catalog No. P08541ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08541ML 1 mL
P08545ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08541ML Supplier TCI America Supplier No. P08541ML

Chemical Identifiers

CAS 20082-71-7
Molecular Formula C8H6ClF5Si
Molecular Weight (g/mol) 260.66
MDL Number MFCD00000500
InChI Key PQRFRTCWNCVQHI-UHFFFAOYSA-N
Synonym pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane
PubChem CID 88361
IUPAC Name chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane
SMILES C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Yellow
Boiling Point 47°C
Quantity 1 mL
UN Number 2985
Formula Weight 260.66
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorophenyldimethylchlorosilane [Pentafluorophenyldimethylsilylating Agent]