missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$76.43 - $76.43
Specifications
| CAS | 1582-24-7 |
|---|---|
| Melting Point | 290°C |
| Color | White |
| Molecular Formula | C6H2BF5O2 |
| MDL Number | MFCD01074663 |
Specifications
| 1582-24-7 | |
| White | |
| MFCD01074663 | |
| (2,3,4,5,6-pentafluorophenyl)boronic acid | |
| 211.88 |
| 290°C | |
| C6H2BF5O2 | |
| VASOMTXTRMYSKD-UHFFFAOYSA-N | |
| 2775922 | |
| Crystalline Powder |