missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P19041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19041G Supplier TCI America Supplier No. P19041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Pentafluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 1582-24-7
Melting Point 290°C
Color White
Molecular Formula C6H2BF5O2
Quantity 1 g
MDL Number MFCD01074663
Synonym 2,3,4,5,6-pentafluorobenzeneboronic acid, pentafluorophenylboronic acid, perfluorophenylboronic acid, pentafluorobenzeneboronic acid, boronic acid, pentafluorophenyl, pentafluorophenyl boronic acid, 2,3,4,5,6-pentafluorophenyl boronic acid, boronic acid, b-2,3,4,5,6-pentafluorophenyl, pubchem22760, rarechem ah pb 0049
InChI Key VASOMTXTRMYSKD-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C(F)=C(F)C(F)=C1F
IUPAC Name (2,3,4,5,6-pentafluorophenyl)boronic acid
Molecular Weight (g/mol) 211.88
PubChem CID 2775922
Formula Weight 211.88
Physical Form Crystalline Powder
Show More Show Less