missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. P10041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10041G Supplier TCI America Supplier No. P10041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 653-21-4
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.102
MDL Number MFCD00004316
InChI Key LGCODSNZJOVMHV-UHFFFAOYSA-N
Synonym pentafluorophenylacetic acid, 2,3,4,5,6-pentafluorophenylacetic acid, pentafluorophenyl acetic acid, 2-perfluorophenyl acetic acid, 2-2,3,4,5,6-pentafluorophenyl acetic acid, 2,3,4,5,6-pentafluorphenylacetic acid, benzeneacetic acid, 2,3,4,5,6-pentafluoro, acmc-20aoj2, pentafluorophenylaceticacid, 2-pentafluorophenyl acetic acid
PubChem CID 69553
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)acetic acid
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Formula Weight 226.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorophenylacetic Acid