missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. p-7135446
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2188200MG 200 mg
P21881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2188200MG Supplier TCI America Supplier No. P2188200MG
Only null left

Chemical Identifiers

CAS 60129-85-3
Molecular Formula C7F8O3S
Molecular Weight (g/mol) 316.121
MDL Number MFCD07784319
InChI Key OTTBRWDUQHDNBY-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid Pentafluorophenyl Ester, Pentafluorophenyl Triflate
PubChem CID 10041425
IUPAC Name (2,3,4,5,6-pentafluorophenyl) trifluoromethanesulfonate
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 40°C
Quantity 200 mg
Formula Weight 316.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorophenyl Trifluoromethanesulfonate