missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl Trifluoroacetate 95.0+%, TCI America™

Catalog No. p-7135308
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18945G 5 g
P189425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18945G Supplier TCI America Supplier No. P18945G
Only null left

Chemical Identifiers

CAS 14533-84-7
Molecular Formula C8F8O2
Molecular Weight (g/mol) 280.07
MDL Number MFCD00134438
InChI Key VCQURUZYYSOUHP-UHFFFAOYSA-N
Synonym pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8
PubChem CID 4327891
IUPAC Name 2,3,4,5,6-pentafluorophenyl 2,2,2-trifluoroacetate
SMILES FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 3272
Formula Weight 280.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorophenyl Trifluoroacetate