missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl trifluoroacetate, 98+%

Catalog No. p-7036266
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5452906 5 g
AAH5452914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5452906 Supplier Thermo Scientific Chemicals Supplier No. H5452906
Only null left

CAS: 14533-84-7 | C8F8O2 | 280.07 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 122.5°Cat760mmHg.

Notes
Hygroscopic. Store at room temperature. Avoid strong oxidizing agents.

Chemical Identifiers

CAS 14533-84-7
Molecular Formula C8F8O2
Molecular Weight (g/mol) 280.07
MDL Number MFCD00134438
InChI Key VCQURUZYYSOUHP-UHFFFAOYSA-N
Synonym pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8
PubChem CID 4327891
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
SMILES FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F

Specifications

Density 1.63
Boiling Point 122°C to 123°C
Flash Point 52°C (126°F)
Refractive Index 1.368
Quantity 5 g
UN Number UN3272
Beilstein 2003848
Sensitivity Moisture sensitive
Solubility Information Soluble in water at 122.5°Cat760mmHg.
Formula Weight 280.07
Percent Purity ≥98%
Chemical Name or Material Pentafluorophenyl trifluoroacetate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only