missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl Benzoate 98.0+%, TCI America™

Catalog No. p-7135470
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22291G 1 g
P22295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22291G Supplier TCI America Supplier No. P22291G
Only null left

Chemical Identifiers

CAS 1548-84-1
Molecular Formula C13H5F5O2
Molecular Weight (g/mol) 288.173
InChI Key WZPWTXZSQHIABL-UHFFFAOYSA-N
Synonym Benzoic Acid Pentafluorophenyl Ester
PubChem CID 11011649
IUPAC Name (2,3,4,5,6-pentafluorophenyl) benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 74°C
Color White
Quantity 1 g
Formula Weight 288.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorophenyl Benzoate