accessibility menu, dialog, popup

UserName

CAS RN 1548-84-1

IUPAC Name: 2,3,4,5,6-pentafluorophenyl benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl benzoate
Molecular Weight (g/mol): 288.17
Molecular Formula: C13H5F5O2
InChi Key: WZPWTXZSQHIABL-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results