missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzyl Bromide 98.0+%, TCI America™
$46.20 - $334.98
Chemical Identifiers
| CAS | 1765-40-8 |
|---|---|
| Molecular Formula | C7H2BrF5 |
| Molecular Weight (g/mol) | 260.989 |
| MDL Number | MFCD00000299 |
| InChI Key | XDEPVFFKOVDUNO-UHFFFAOYSA-N |
| Synonym | 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide |
| PubChem CID | 74484 |
| IUPAC Name | 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene |
| SMILES | C(C1=C(C(=C(C(=C1F)F)F)F)F)Br |
Chemical Identifiers
| 1765-40-8 | |
| 260.989 | |
| XDEPVFFKOVDUNO-UHFFFAOYSA-N | |
| 74484 | |
| C(C1=C(C(=C(C(=C1F)F)F)F)F)Br |
| C7H2BrF5 | |
| MFCD00000299 | |
| 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide | |
| 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene |
Specifications
| 1765-40-8 | |
| 176°C | |
| MFCD00000299 | |
| 3265 | |
| XDEPVFFKOVDUNO-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene | |
| 74484 | |
| ≥98.0% (GC) | |
| Pentafluorobenzyl Bromide |
| Yellow | |
| C7H2BrF5 | |
| 1 g | |
| 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide | |
| C(C1=C(C(=C(C(=C1F)F)F)F)F)Br | |
| 260.989 | |
| 260.99 | |
| Liquid |
Safety and Handling
TSCA : No