missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. P08095G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08095G 5 g
P08091G 1 g
P080925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P08095G Supplier TCI America Supplier No. P08095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1765-40-8
Molecular Formula C7H2BrF5
Molecular Weight (g/mol) 260.989
MDL Number MFCD00000299
InChI Key XDEPVFFKOVDUNO-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
PubChem CID 74484
IUPAC Name 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)Br

Specifications

Color Yellow
Boiling Point 176°C
Quantity 5 g
UN Number 3265
Formula Weight 260.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorobenzyl Bromide