missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. P08075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08075G 5 g
P080725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08075G Supplier TCI America Supplier No. P08075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2251-50-5
Molecular Formula C7ClF5O
Molecular Weight (g/mol) 230.52
MDL Number MFCD00000657
InChI Key MYHOHFDYWMPGJY-UHFFFAOYSA-N
Synonym pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
PubChem CID 75256
ChEBI CHEBI:39425
IUPAC Name pentafluorobenzoyl chloride
SMILES FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F

Specifications

Color Yellow
Boiling Point 159°C
Quantity 5 g
UN Number 3265
Formula Weight 230.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Pentafluorobenzoyl Chloride