missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl chloride, 98%

Catalog No. AAA1242022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1242022 100 g
AAA1242006 5 g
AAA1242014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1242022 Supplier Thermo Scientific Chemicals Supplier No. A1242022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2251-50-5 | C7ClF5O | 230.52 g/mol

A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Water, Alcohols, Strong bases.

Chemical Identifiers

CAS 2251-50-5
Molecular Formula C7ClF5O
Molecular Weight (g/mol) 230.52
MDL Number MFCD00000657
InChI Key MYHOHFDYWMPGJY-UHFFFAOYSA-N
Synonym pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
PubChem CID 75256
ChEBI CHEBI:39425
IUPAC Name 2,3,4,5,6-pentafluorobenzoyl chloride
SMILES FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F

Specifications

Density 1.668
Boiling Point 158°C to 159°C
Refractive Index 1.4535
Quantity 100 g
UN Number UN3265
Beilstein 2054397
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 230.52
Percent Purity 98%
Chemical Name or Material Pentafluorobenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-842-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only