missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzonitrile 98.0+%, TCI America™
$44.28 - $103.97
Chemical Identifiers
| CAS | 773-82-0 |
|---|---|
| Molecular Formula | C7F5N |
| Molecular Weight (g/mol) | 193.076 |
| MDL Number | MFCD00001775 |
| InChI Key | YXWJGZQOGXGSSC-UHFFFAOYSA-N |
| Synonym | pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k |
| PubChem CID | 69882 |
| IUPAC Name | 2,3,4,5,6-pentafluorobenzonitrile |
| SMILES | C(#N)C1=C(C(=C(C(=C1F)F)F)F)F |
Chemical Identifiers
| 773-82-0 | |
| 193.076 | |
| YXWJGZQOGXGSSC-UHFFFAOYSA-N | |
| 69882 | |
| C(#N)C1=C(C(=C(C(=C1F)F)F)F)F |
| C7F5N | |
| MFCD00001775 | |
| pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k | |
| 2,3,4,5,6-pentafluorobenzonitrile |
Specifications
| 773-82-0 | |
| 162°C | |
| MFCD00001775 | |
| 1993 | |
| YXWJGZQOGXGSSC-UHFFFAOYSA-N | |
| 2,3,4,5,6-pentafluorobenzonitrile | |
| 69882 | |
| ≥98.0% (GC) | |
| Pentafluorobenzonitrile |
| Colorless | |
| C7F5N | |
| 5 g | |
| pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k | |
| C(#N)C1=C(C(=C(C(=C1F)F)F)F)F | |
| 193.076 | |
| 193.08 | |
| Liquid |
Safety and Handling
TSCA : Yes