missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzonitrile 98.0+%, TCI America™

Catalog No. P09355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09355G 5 g
P093525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09355G Supplier TCI America Supplier No. P09355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 773-82-0
Molecular Formula C7F5N
Molecular Weight (g/mol) 193.076
MDL Number MFCD00001775
InChI Key YXWJGZQOGXGSSC-UHFFFAOYSA-N
Synonym pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k
PubChem CID 69882
IUPAC Name 2,3,4,5,6-pentafluorobenzonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color Colorless
Boiling Point 162°C
Quantity 5 g
UN Number 1993
Formula Weight 193.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorobenzonitrile