missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzonitrile, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 773-82-0 |
|---|---|
| Molecular Formula | C7F5N |
| Molecular Weight (g/mol) | 193.07 |
| InChI Key | YXWJGZQOGXGSSC-UHFFFAOYSA-N |
| Synonym | pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k |
| PubChem CID | 69882 |
| IUPAC Name | 2,3,4,5,6-pentafluorobenzonitrile |
| SMILES | C(#N)C1=C(C(=C(C(=C1F)F)F)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC188340050
|
Thermo Scientific Chemicals
188340050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC188340250
|
Thermo Scientific Chemicals
188340250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC188341000
|
Thermo Scientific Chemicals
188341000 |
100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 773-82-0 | |
| 193.07 | |
| pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k | |
| 2,3,4,5,6-pentafluorobenzonitrile |
| C7F5N | |
| YXWJGZQOGXGSSC-UHFFFAOYSA-N | |
| 69882 | |
| C(#N)C1=C(C(=C(C(=C1F)F)F)F)F |
Specifications
| 773-82-0 | |
| 161.0°C to 162.0°C | |
| Authentic | |
| C7F5N | |
| C6F5CN | |
| 1.53 | |
| Solubility in water: insoluble. | |
| C(#N)C1=C(C(=C(C(=C1F)F)F)F)F | |
| 193.07 | |
| 193.07 | |
| Pentafluorobenzonitrile |
| 1.5300g/mL | |
| 29°C | |
| 98.5% min. (GC) | |
| 1.4415 to 1.4435 | |
| 5g | |
| pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k | |
| YXWJGZQOGXGSSC-UHFFFAOYSA-N | |
| 2,3,4,5,6-pentafluorobenzonitrile | |
| 69882 | |
| 99% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 212-259-9
RUO â Research Use Only