missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzonitrile, 98%

Catalog No. AAA1357522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1357522 100 g
AAA1357506 5 g
AAA1357514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1357522 Supplier Thermo Scientific Chemicals Supplier No. A1357522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 773-82-0 | C7F5N | 193.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 773-82-0
Molecular Formula C7F5N
Molecular Weight (g/mol) 193.076
MDL Number MFCD00001775
InChI Key YXWJGZQOGXGSSC-UHFFFAOYSA-N
Synonym pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k
PubChem CID 69882
IUPAC Name 2,3,4,5,6-pentafluorobenzonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 1°C to 2°C
Density 1.566
Boiling Point 161°C to 162°C
Flash Point 29°C (84°F)
Refractive Index 1.4425
Quantity 100 g
UN Number UN1992
Beilstein 1913453
Formula Weight 193.07
Percent Purity 98%
Chemical Name or Material Pentafluorobenzonitrile
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 212-259-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only