missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoic Acid 98.0+%, TCI America™

Catalog No. P08065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08065G 5 g
P080625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08065G Supplier TCI America Supplier No. P08065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 602-94-8
Molecular Formula C7HF5O2
Molecular Weight (g/mol) 212.08
MDL Number MFCD00002406
InChI Key YZERDTREOUSUHF-UHFFFAOYSA-N
Synonym pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017
PubChem CID 11770
ChEBI CHEBI:46796
IUPAC Name pentafluorobenzoic acid
SMILES OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 212.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorobenzoic Acid
EINECSNumber (3)-3800
RTECSNumber DH6195000
TSCA Yes