missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoic acid, 98+%

CAS: 602-94-8 | C7HF5O2 | 212.08 g/mol

$77.32 - $263.03

Chemical Identifiers

CAS 602-94-8
Molecular Formula C7HF5O2
Molecular Weight (g/mol) 212.08
MDL Number MFCD00002406
InChI Key YZERDTREOUSUHF-UHFFFAOYSA-N
Synonym pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017
PubChem CID 11770
ChEBI CHEBI:46796
IUPAC Name 2,3,4,5,6-pentafluorobenzoic acid
SMILES OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1239606
View Documents
Thermo Scientific Chemicals
A1239606
5 g
Each for $77.32
Only null left
 
AAA1239614
View Documents
Thermo Scientific Chemicals
A1239614
25 g
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

602-94-8
212.08
YZERDTREOUSUHF-UHFFFAOYSA-N
11770
2,3,4,5,6-pentafluorobenzoic acid
C7HF5O2
MFCD00002406
pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017
CHEBI:46796
OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

602-94-8
219°C to 221°C
MFCD00002406
2054395
YZERDTREOUSUHF-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzoic acid
11770
212.07
Pentafluorobenzoic acid
100°C to 104°C
C7HF5O2
5 g
pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017
OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
212.08
CHEBI:46796
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-026-6

RTECSNumber : DH6195000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only