missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzenethiol 98.0+%, TCI America™

Catalog No. p-7134816
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
P08615G 5 g
P086125G 25 g
2 options

Catalog No. P08615G

Supplier: TCI America P08615G

Only null left

Chemical Identifiers

CAS 771-62-0
Molecular Formula C6HF5S
Molecular Weight (g/mol) 200.13
MDL Number MFCD00004828
InChI Key UVAMFBJPMUMURT-UHFFFAOYSA-N
Synonym pentafluorothiophenol, pentafluorobenzenethiol, benzenethiol, pentafluoro, pentafluorophenyl mercaptan, 2,3,4,5,6-pentafluorothiophenol, 2,3,4,5,6-pentafluorobenzene-1-thiol, pentafluorobenzene thiol, pentafluorobenzene-1-thiol, 2,3,4,5,6-pentafluorothiphenol
PubChem CID 13042
IUPAC Name pentafluorobenzene-1-thiol
SMILES FC1=C(F)C(F)=C(S)C(F)=C1F

Specifications

Melting Point -24°C
Color Yellow
Boiling Point 143°C
Quantity 5 g
UN Number 2920
Formula Weight 200.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorobenzenethiol
RTECSNumber DC1940000
TSCA No
Recommended Storage Refrigerator