missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America P093425G
Specifications
| Pentafluorobenzenesulfonyl Chloride | |
| Yellow | |
| C6ClF5O2S | |
| 25 g | |
| pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride | |
| C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F | |
| 266.566 | |
| 266.57 | |
| Liquid |
| 832-53-1 | |
| 210°C | |
| MFCD00007427 | |
| 3265 | |
| UOJCTEGNHXRPKO-UHFFFAOYSA-N | |
| 2,3,4,5,6-pentafluorobenzenesulfonyl chloride | |
| 70026 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 832-53-1 | |
| 266.566 | |
| UOJCTEGNHXRPKO-UHFFFAOYSA-N | |
| 70026 | |
| C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F |
| C6ClF5O2S | |
| MFCD00007427 | |
| pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride | |
| 2,3,4,5,6-pentafluorobenzenesulfonyl chloride |
Safety and Handling
TSCA : No